Search
Search

Gm15455| Gene Symbol: Gm15455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161816.1 | NP_001155288.1 | 1 | 2830 | 129 | |
| GenBank mRNA | AK139531.1 | - | 1601 | 129 | ||
| BC157984.1 | - | 1 | 2754 | 129 | ||
| BC157993.1 | - | 1 | 2754 | 129 | ||
Set Membership: |
|