| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00000720.mm.1 |
NM_199063.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000720.mm.1 |
NM_199063.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000720.mm.1 |
TC0X00003386.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000723.mm.1 |
NM_199063.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000723.mm.1 |
NM_199063.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000723.mm.1 |
TC0X00003386.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000721.mm.1 |
NM_199063.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000721.mm.1 |
NM_199063.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000721.mm.1 |
TC0X00003386.mm.1 |
MTA TC |