Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01610603_gH
          See other PISD-PS1 GE Assays ›
          Gene Symbol:
          Pisd-ps1
          Gene Name:
          phosphatidylserine decarboxylase, pseudogene 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 3124021 - 3131945 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01610603_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pisd-ps1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003517.1 3 - 4 543 162
          GenBank mRNA CX562509.1 - 2 - 3 397 162

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000001.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049211 NR_003517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049211 AI481313.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133278 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049211 NR_003517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049211 AI481313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154668 NR_003517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154668 AI481313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154668 BC059938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154668 BC034366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154668 BC048401.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373704 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373708 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373702 - -
          Mouse Genome 430 2.0 Array 1454566_at - -
          Mouse Genome 430 2.0 Array 1439070_PM_x_at - -
          Mouse Genome 430 2.0 Array 1439070_x_at - -
          Mouse Genome 430 2.0 Array 1436944_PM_x_at - -
          Mouse Genome 430 2.0 Array 1434975_PM_x_at - -
          Mouse Genome 430 2.0 Array 1434975_x_at - -
          Mouse Genome 430 2.0 Array 1453145_PM_at - -
          Mouse Genome 430 2.0 Array 1435353_PM_a_at - -
          Mouse Genome 430 2.0 Array 1436944_x_at - -
          Mouse Genome 430 2.0 Array 1453144_PM_at - -
          Mouse Genome 430 2.0 Array 1435426_PM_s_at - -
          Mouse Genome 430 2.0 Array 1441414_at - -
          Mouse Genome 430 2.0 Array 1426387_PM_x_at - -
          Mouse Genome 430 2.0 Array 1453144_at - -
          Mouse Genome 430 2.0 Array 1429452_PM_x_at - -
          Mouse Genome 430 2.0 Array 1435425_PM_at - -
          Mouse Genome 430 2.0 Array 1435426_s_at - -
          Mouse Genome 430 2.0 Array 1453145_at - -
          Mouse Genome 430 2.0 Array 1441414_PM_at - -
          Mouse Genome 430 2.0 Array 1429452_x_at - -
          Mouse Genome 430 2.0 Array 1435425_at - -
          Mouse Genome 430 2.0 Array 1435353_a_at - -
          Mouse Genome 430 2.0 Array 1426387_x_at - -
          Mouse Genome 430 2.0 Array 1454566_PM_at - -
          Mouse Genome 430 2.0 Array 1439069_a_at - -
          Mouse Genome 430 2.0 Array 1439069_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 236604_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 236604_at - -
          Affymetrix Mouse Clariom S Array PSR1100000010.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000010.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000034.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000034.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000009.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000009.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000021.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000021.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000002.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000002.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000005.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000005.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000029.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000029.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000033.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000033.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000022.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000022.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000027.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000027.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000017.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000017.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000007.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000007.mm.1 TC1100000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000023.mm.1 NR_003517.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000023.mm.1 TC1100000001.mm.1 MTA TC
          Mouse Expression Set 430 1435353_a_at - -
          Mouse Expression Set 430 1436944_x_at - -
          Mouse Expression Set 430 1426387_PM_x_at - -
          Mouse Expression Set 430 1439070_PM_x_at - -
          Mouse Expression Set 430 1439070_x_at - -
          Mouse Expression Set 430 1435353_PM_a_at - -
          Mouse Expression Set 430 1426387_x_at - -
          Mouse Expression Set 430 1436944_PM_x_at - -
          Mouse Expression Set 430 1454566_PM_at - -
          Mouse Expression Set 430 1454566_at - -
          Murine Genome U74v2 Set 97448_at - -
          Murine Genome U74v2 Set 97448_PM_at - -
          Mouse Genome 430A 2.0 Array 1435353_a_at - -
          Mouse Genome 430A 2.0 Array 1426387_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1436944_x_at - -
          Mouse Genome 430A 2.0 Array 1439070_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1426387_x_at - -
          Mouse Genome 430A 2.0 Array 1439070_x_at - -
          Mouse Genome 430A 2.0 Array 1435353_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1436944_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2944601 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline