Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01610865_sH
          See other OLFR10 GE Assays ›
          Gene Symbol:
          Olfr10
          Gene Name:
          olfactory receptor 10
          Gene Aliases:
          GA_x6K02T2QP88-6117098-6116163, L45, MOR256-55
          Chromosome Location:
          Chr.11: 49317548 - 49318483 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01610865_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206822.1 NP_996558.1 1 666 155
          GenBank mRNA BC148303.2 - 1 666 40041401 155

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr10

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989608 NM_206822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989608 BC148303.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989608 NM_206822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989608 BC148303.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09665596 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09665582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782494 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375548 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375547 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 870070 - -
          Illumina mouse-6 v1.0 expression beadchip 870070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870070 - -
          mouse-6 v1.1 (Illumina) scl018307.1_307-S - -
          Illumina Sentrix mouse-6.v1 bead array 870070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018307.1_307-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3003383 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2701018 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3003383 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline