Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01611149_sH
          See other OLFR804 GE Assays ›
          Gene Symbol:
          Olfr804
          Gene Name:
          olfactory receptor 804
          Gene Aliases:
          MOR110-7
          Chromosome Location:
          Chr.10: 129704880 - 129705824 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01611149_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr804
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011821.1 NP_001011821.1 1 241 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr804

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110335 NM_001011821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110335 NM_001011821.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546927 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118177 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107969 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121973 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112929 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08275594 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08955704 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08275605 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08955712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08619534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08275602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546890 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367490 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367489 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367483 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367517 - -
          Affymetrix Mouse Clariom S Array PSR1000012050.mm.1 NM_001011821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012050.mm.1 NM_001011821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012050.mm.1 TC1000001652.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161495 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161495 - -
          mouse-6 v1.1 (Illumina) GI_58801405-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161495 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161495 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline