Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01611369_s1
          See other OLFR118 GE Assays ›
          Gene Symbol:
          Olfr118
          Gene Name:
          olfactory receptor 118
          Gene Aliases:
          MOR263-13
          Chromosome Location:
          Chr.17: 37672025 - 37672990 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01611369_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr118
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_213721.2 NP_998886.2 1 552 101
          GenBank mRNA BC128012.1 - 1 556 40136025 101

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr118

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040205 NM_213721.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040205 BC128012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040205 NM_213721.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040205 BC128012.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169544 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134102 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140200 NM_146631.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116423 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13483901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13483899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13554122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13496521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13496512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13062181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169532 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445146 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445145 - -
          Affymetrix Mouse Clariom S Array PSR1700007727.mm.1 NM_213721.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007727.mm.1 NM_213721.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007727.mm.1 TC1700000778.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 633986 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 110463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230068 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230068 - -
          mouse-6 v1.1 (Illumina) scl00404308.1_197-S - -
          Illumina Sentrix mouse-6.v1 bead array 110463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00404308.1_197-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2663239 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2663239 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline