Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01611922_sH
          See other OLFR1306 GE Assays ›
          Gene Symbol:
          Olfr1306
          Gene Name:
          olfactory receptor 1306
          Gene Aliases:
          MOR245-15
          Chromosome Location:
          Chr.2: 111911990 - 111912928 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01611922_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1306
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011803.2 NP_001011803.2 1 735 130

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1306

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285731 NM_001011803.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285731 NM_001011803.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285731 NM_001011803.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285731 NM_001011803.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285731 NM_001011803.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285731 NM_001011803.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285731 NM_001011803.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285731 NM_001011803.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01676216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01531147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526714 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485797 - -
          Affymetrix Mouse Gene 1.1 ST Array 10485796 - -
          Affymetrix Mouse Clariom S Array PSR0200036236.mm.1 NM_001011803.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036236.mm.1 NM_001011803.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036236.mm.1 TC0200004334.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162277 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162277 - -
          mouse-6 v1.1 (Illumina) GI_58801371-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162277 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162277 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline