Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01612956_gH
          See other NR2C2AP GE Assays ›
          Gene Symbol:
          Nr2c2ap
          Gene Name:
          nuclear receptor 2C2-associated protein
          Gene Aliases:
          2310073E15Rik
          Chromosome Location:
          Chr.8: 70131327 - 70133752 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01612956_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr2c2ap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025586.2 NP_001020757.1 2 - 3 367 150
          NM_001025587.2 NP_001020758.1 2 - 3 367 150
          GenBank mRNA AK160346.1 - 2 - 3 348 150
          BC092288.1 - 2 - 3 311 5003230 150
          BY722132.1 - 2 - 3 336 150

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000737.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr2c2ap

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25950004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25950003 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25950005 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 AK010864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 AK160346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 AK168268.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 BC092288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 NM_001025586.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 AK010864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 AK160346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 AK168268.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 BC092288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314153 NM_001025586.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 AK010864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 AK160346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 AK168268.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 BC092288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 NM_001025586.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 AK010864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 AK160346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 AK168268.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 BC092288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314153 NM_001025586.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25950025 - -
          Whole Mouse Genome G4122A A_51_P314153 AK010864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 AK160346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 AK168268.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 BC092288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 NM_001025586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 AK010864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 AK160346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 AK168268.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 BC092288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314153 NM_001025586.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109592 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07427079 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07427090 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07427085 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25950003 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25950005 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25950004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25949972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25949976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25949978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25950016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25950018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25950005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25950014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25950024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25950003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07427093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25950006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07399491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07427084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07427081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25950026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25950016 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10579188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579187 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579182 - -
          Affymetrix HT MG-430 PM Array Plate 1447876_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1428534_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10579181 - -
          Affymetrix Mouse Gene 1.1 ST Array 10572290 - -
          Mouse Genome 430 2.0 Array 1428534_at - -
          Mouse Genome 430 2.0 Array 1447876_PM_x_at - -
          Mouse Genome 430 2.0 Array 1428534_PM_at - -
          Mouse Genome 430 2.0 Array 1447876_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75692_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75692_at - -
          Affymetrix Mouse Clariom S Array PSR0800005047.mm.1 NM_001025586.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005047.mm.1 NM_001025587.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005047.mm.1 TC0800000737.mm.1 MTA TC
          Mouse Expression Set 430 1428534_PM_at - -
          Mouse Expression Set 430 1428534_at - -
          Mouse Expression Set 430 1447876_PM_x_at - -
          Mouse Expression Set 430 1447876_x_at - -
          Murine Genome U74v2 Set 161841_PM_r_at - -
          Murine Genome U74v2 Set 160654_PM_at - -
          Murine Genome U74v2 Set 161841_r_at - -
          Murine Genome U74v2 Set 160654_at - -
          Murine Genome U74v2 Set 137162_at - -
          Murine Genome U74v2 Set 137162_PM_at - -
          Mouse Genome 430A 2.0 Array 1428534_PM_at - -
          Mouse Genome 430A 2.0 Array 1428534_at - -
          Affymetrix HT Mouse Genome 430A Array 1428534_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428534_at - -
          Affymetrix HT Mouse Genome 430B Array 1447876_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447876_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233032 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3084818 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3084818 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline