Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01613158_m1
          See other GBP11 GE Assays ›
          Gene Symbol:
          Gbp11
          Gene Name:
          guanylate binding protein 11
          Gene Aliases:
          EG634650, Gm7141
          Chromosome Location:
          Chr.5: 105323026 - 105346472 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01613158_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gbp11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039647.2 NP_001034736.2 5 - 6 703 128
          GenBank mRNA BC111039.1 - 5 - 6 700 5349539 128
          EU304258.1 - 4 - 5 620 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002896.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042813 BC111039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042813 BI655333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042813 NM_001039647.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28738 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7739 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042813 BC111039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042813 BI655333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042813 NM_001039647.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23954270 - -
          Whole Mouse Genome G4122A A_51_P327405 BC080665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327405 BC031475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327405 NM_029509.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327405 AK017988.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23954738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04811886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23954953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04905899 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532000 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532017 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532018 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532021 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531998 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532009 - -
          Affymetrix Mouse Gene 1.1 ST Array 10531994 - -
          Affymetrix Mouse Gene 1.1 ST Array 10532019 - -
          Affymetrix Mouse Clariom S Array PSR0500025640.mm.1 TC0500002896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025639.mm.1 TC0500002896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025643.mm.1 TC0500002896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025642.mm.1 TC0500002896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025641.mm.1 TC0500002896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025644.mm.1 TC0500002896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025646.mm.1 TC0500002896.mm.1 MTA TC
          Mouse Expression Set 430 1421104_PM_at - -
          Mouse Expression Set 430 1421104_at - -
          Mouse Expression Set 430 1447927_at - -
          Mouse Expression Set 430 1447927_PM_at - -
          Murine Genome U74v2 Set 160523_at - -
          Murine Genome U74v2 Set 160523_PM_at - -
          Mouse Genome 430A 2.0 Array 1421104_PM_at - -
          Mouse Genome 430A 2.0 Array 1421104_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242854 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2837493 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2837493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline