Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01613327_g1
          See other PSAT1 GE Assays ›
          Gene Symbol:
          Psat1
          Gene Name:
          phosphoserine aminotransferase 1
          Gene Aliases:
          D8Ertd814e, EPIP, PSA, Psat
          Chromosome Location:
          Chr.19: 15905123 - 15925059 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01613327_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177420.2 NP_803155.1 1 - 2 197 107
          GenBank mRNA AF259674.1 - 1 - 2 95 107
          AK032487.1 - 1 - 2 198 107
          AK035361.1 - 1 - 2 87 107
          AK049855.1 - 1 - 2 131 107
          AK090108.1 - 1 - 2 125 107
          AK145216.1 - 1 - 2 113 107
          AK145672.1 - 1 - 2 122 107
          AK162609.1 - 1 - 2 88 107
          AK166880.1 - 1 - 2 122 107
          AK167029.1 - 1 - 2 88 107
          AK167044.1 - 1 - 2 135 107
          AK168098.1 - 1 - 2 112 107
          AK168216.1 - 1 - 2 135 107
          AK169370.1 - 1 - 2 112 107
          BC004827.1 - 1 - 2 163 2616298 107

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF259674.1, AK049855.1, AK090108.1, AK145216.1, AK145672.1, AK162609.1, AK166880.1, AK167029.1, AK167044.1, AK168098.1, AK169370.1, BC004827.1, NM_177420.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psat1

          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          L-serine biosynthetic process
          metabolic process
          cellular amino acid biosynthetic process
          catalytic activity
          O-phospho-L-serine:2-oxoglutarate aminotransferase activity
          transaminase activity
          transferase activity
          pyridoxal phosphate binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900010958.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010958.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010958.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010957.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010957.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010957.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010956.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010956.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010956.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010953.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010953.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010953.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010953.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010953.mm.1 TC1900001203.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893894 - -
          ABI Mouse Genome Survey Microarray 916661 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline