Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01614382_m1
          See other CLCA4A GE Assays ›
          Gene Symbol:
          Clca4a
          Gene Name:
          chloride channel calcium activated 4A
          Gene Aliases:
          9130020L07Rik, AI504701, Clca4, Clca6
          Chromosome Location:
          Chr.3: 144952486 - 144975045 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01614382_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clca4a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207208.3 NP_997091.3 6 - 7 995 138
          GenBank mRNA AK033591.1 - 6 - 7 995 138
          AY560902.2 - 961 138
          AY560903.2 - 961 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003083.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clca4a

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          transport
          chloride transport
          chloride transmembrane transport
          intracellular calcium activated chloride channel activity
          chloride channel activity
          peptidase activity
          cysteine-type peptidase activity
          metallopeptidase activity
          ligand-gated ion channel activity
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982284 AK033591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982284 NM_207208.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 95137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41083 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126019 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982284 AK033591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982284 NM_207208.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P134993 AK054018.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P134993 AK033591.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P134993 NM_207208.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605904 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105508 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105492 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126019 - -
          Whole Mouse Genome G4122A A_52_P95161 BC139088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95161 BC139089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95161 NM_001033199.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95161 AK136515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95161 BC139088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95161 BC139089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95161 NM_001033199.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95161 AK136515.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133110 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137359 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114302 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111487 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126019 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05105492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02964826 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02964852 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02964856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02768070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02376837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02430799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02887110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02430790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03113506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02376836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03177343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03113494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02755937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02964826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605915 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10502605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502600 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502597 - -
          Affymetrix HT MG-430 PM Array Plate 1439727_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10502606 - -
          Affymetrix Mouse Gene 1.1 ST Array 10502590 - -
          Mouse Genome 430 2.0 Array 1443256_at - -
          Mouse Genome 430 2.0 Array 1447394_at - -
          Mouse Genome 430 2.0 Array 1439727_PM_at - -
          Mouse Genome 430 2.0 Array 1447394_PM_at - -
          Mouse Genome 430 2.0 Array 1439727_at - -
          Mouse Genome 430 2.0 Array 1443256_PM_at - -
          Mouse Genome 430 2.0 Array 1439796_PM_at - -
          Mouse Genome 430 2.0 Array 1439796_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 99663_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 99663_at - -
          Affymetrix Mouse Clariom S Array PSR0300024255.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024255.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024255.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024264.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024264.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024264.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024250.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024250.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024250.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024262.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024262.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024262.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024266.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024266.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024266.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024257.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024257.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024257.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024260.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024260.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024260.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024254.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024254.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024254.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024256.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024256.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024256.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024261.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024261.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024261.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024265.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024265.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024265.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024268.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024268.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024268.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024253.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024253.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024253.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024252.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024252.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024252.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024258.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024258.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024258.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024259.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024259.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024259.mm.1 TC0300003083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024263.mm.1 NM_207208.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024263.mm.1 NM_207208.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024263.mm.1 TC0300003083.mm.1 MTA TC
          Mouse Expression Set 430 1439727_at - -
          Mouse Expression Set 430 1439727_PM_at - -
          Mouse Expression Set 430 1447394_at - -
          Mouse Expression Set 430 1447394_PM_at - -
          Murine Genome U74v2 Set 166879_PM_at - -
          Murine Genome U74v2 Set 166879_at - -
          Affymetrix HT Mouse Genome 430B Array 1439727_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439727_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2742695 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2752464 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659889 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2742695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659889 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2752464 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0