Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01614978_sH
          See other OLFR458 GE Assays ›
          Gene Symbol:
          Olfr458
          Gene Name:
          olfactory receptor 458
          Gene Aliases:
          MOR211-8P, MOR221-4, MOR257-4, Olfr301
          Chromosome Location:
          Chr.6: 42460076 - 42461017 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01614978_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr458
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146444.1 NP_666655.1 1 365 152
          GenBank mRNA BC106796.1 - 1 365 152
          BC106797.1 - 1 365 152
          BC119415.1 - 1 365 152
          BC119416.1 - 1 365 152
          BC127219.1 - 1 360 152
          BC127220.1 - 1 365 152

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr458

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169046 NM_146444.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141805 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169046 NM_146444.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589468 BC127219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589468 BC127220.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589468 BC119416.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589468 NM_146444.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589468 BC106796.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398462 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141805 - -
          Whole Mouse Genome G4122A A_52_P589468 BC127219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P589468 BC127220.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P589468 BC119416.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P589468 NM_146444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P589468 BC106796.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100354 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141805 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05029519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105932 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05105921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05157686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05029503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05803763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398479 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10544439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544441 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544436 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544438 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544440 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5890524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005890524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232841 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232841 - -
          mouse-6 v1.1 (Illumina) scl29078.1.1_230-S - -
          Illumina Sentrix mouse-6.v1 bead array 5890524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29078.1.1_230-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2925730 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232841 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2925730 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline