| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10441861 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10441860 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10526719 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10441859 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10526721 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10526718 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10526720 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10441858 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1700001200.mm.1 |
NM_013741.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001200.mm.1 |
NM_013741.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001200.mm.1 |
TC1700000152.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001199.mm.1 |
NM_013741.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001199.mm.1 |
NM_013741.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001199.mm.1 |
TC1700000152.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001201.mm.1 |
NM_013741.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001201.mm.1 |
NM_013741.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001201.mm.1 |
TC1700000152.mm.1 |
MTA TC |