Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sult2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101586.2 | NP_001095056.2 | 5 - 6 | 749 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus 1700 Inventoried Probe spans exons |