Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01615387_s1
          See other OLFR1259 GE Assays ›
          Gene Symbol:
          Olfr1259
          Gene Name:
          olfactory receptor 1259
          Gene Aliases:
          MOR232-9
          Chromosome Location:
          Chr.2: 89943184 - 89944113 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01615387_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1259
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146341.1 NP_666453.1 1 774 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1259

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968608 NM_146341.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162956 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968608 NM_146341.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162956 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01312230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01354413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01515135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01312233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01995011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02086997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01994997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415236 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484857 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484856 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3990400 - -
          Illumina mouse-6 v1.0 expression beadchip 3990400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230644 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230644 - -
          mouse-6 v1.1 (Illumina) scl00258338.1_328-S - -
          Illumina Sentrix mouse-6.v1 bead array 3990400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258338.1_328-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230644 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230644 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline