Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01615657_s1
          See other OLFR301 GE Assays ›
          Gene Symbol:
          Olfr301
          Gene Name:
          olfactory receptor 301
          Gene Aliases:
          MOR211-8P, MOR221-1P, MOR221-4, Olfr1531-ps1
          Chromosome Location:
          Chr.7: 86403849 - 86413299 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01615657_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr301
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_212436.2 NP_997601.1 3 827 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr301

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098156 NM_212436.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727043 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098156 NM_212436.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294630 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06176774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06176775 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06176768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554722 - -
          Affymetrix Mouse Gene 1.1 ST Array 10554721 - -
          Affymetrix Mouse Clariom S Array PSR0700010766.mm.1 TC0700001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010766.mm.1 NM_212436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010766.mm.1 NM_212436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010767.mm.1 TC0700001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010767.mm.1 NM_212436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010767.mm.1 NM_212436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010768.mm.1 TC0700001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010768.mm.1 NM_212436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010768.mm.1 NM_212436.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 375333 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106450161 - -
          Illumina mouse-6 v1.0 expression beadchip 106450161 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2652144 - -
          mouse-6 v1.1 (Illumina) scl0257958.1_223-S - -
          Illumina Sentrix mouse-6.v1 bead array 106450161 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106450161 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2919904 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2652144 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2919904 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0