Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01615926_m1
          See other UGT2B36 GE Assays ›
          Gene Symbol:
          Ugt2b36
          Gene Name:
          UDP glucuronosyltransferase 2 family, polypeptide B36
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 87065927 - 87092555 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01615926_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt2b36
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029867.1 NP_001025038.1 5 - 6 1346 156
          GenBank mRNA AK149425.1 - 5 - 6 1349 156
          BC141232.1 - 5 - 6 1341 9055844 156
          BC145555.1 - 5 - 6 1275 9054119 156

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002692.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugt2b36

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          flavonoid biosynthetic process
          cellular response to hormone stimulus
          flavonoid glucuronidation
          glucuronosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P102438 BC145555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P102438 NM_001029867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P102438 AK149425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P102438 BC141232.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P102438 BC145555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P102438 NM_001029867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P102438 AK149425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P102438 BC141232.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125888 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56974 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39280 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102438 BC145555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102438 NM_001029867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102438 AK149425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102438 BC141232.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102438 BC145555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102438 NM_001029867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102438 AK149425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102438 BC141232.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23866624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23866616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23866615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23866634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23866630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23866629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23866637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23866640 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05644259 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05644234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05644226 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05644238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23866489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04559781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04559093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04405071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04206370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04559087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04678083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04704695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04937431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04206364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23866634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10531052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531053 - -
          Affymetrix HT MG-430 PM Array Plate 1455595_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10531051 - -
          Mouse Genome 430 2.0 Array 1455595_PM_at - -
          Mouse Genome 430 2.0 Array 1455595_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231396_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231396_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500023756.mm.1 NM_001029867.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023756.mm.1 NM_001029867.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023756.mm.1 TC0500002692.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023754.mm.1 NM_001029867.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023754.mm.1 NM_001029867.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023754.mm.1 TC0500002692.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023753.mm.1 NM_001029867.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023753.mm.1 NM_001029867.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023753.mm.1 TC0500002692.mm.1 MTA TC
          Mouse Expression Set 430 1455595_PM_at - -
          Mouse Expression Set 430 1455595_at - -
          Affymetrix HT Mouse Genome 430B Array 1455595_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455595_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 490672 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235719 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2955818 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2955818 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline