Search
Search

Olfr154| Gene Symbol: Olfr154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013728.2 | NP_038756.2 | 1 | 466 | 154 | |
| GenBank mRNA | BC120642.1 | - | 1 | 466 | 40129565 | 154 |
| BC120644.1 | - | 1 | 466 | 40129567 | 154 | |
Set Membership: |
|