Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01615963_g1
          See other COX11 GE Assays ›
          Gene Symbol:
          Cox11
          Gene Name:
          cytochrome c oxidase assembly protein 11
          Gene Aliases:
          2010004I09Rik
          Chromosome Location:
          Chr.11: 90638184 - 90645977 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01615963_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199008.2 NP_950173.1 2 - 3 526 106
          GenBank mRNA AK008096.1 - 2 - 3 540 106
          AK143937.1 - 2 - 3 526 106
          AK172067.1 - 2 - 3 530 106
          BC061233.1 - 2 - 3 537 6585499 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox11

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of glucokinase activity
          copper ion binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28013001 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28013004 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 AK083630.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 BC061233.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 AK143937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 AK172067.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 AK008096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 AK083630.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 BC061233.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 AK143937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 AK172067.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214319 AK008096.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162115 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879977 AF229257.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879977 AK038825.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879977 NM_021323.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67552 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151319 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165083 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 AK083630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 BC061233.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 AK143937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 AK172067.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 AK008096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 AK083630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 BC061233.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 AK143937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 AK172067.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214319 AK008096.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 AK083630.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 BC061233.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 AK143937.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 AK172067.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 AK008096.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 AK083630.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 BC061233.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 AK143937.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 AK172067.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P214319 AK008096.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28013005 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151319 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165083 - -
          Whole Mouse Genome G4122A A_51_P214319 AK083630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 BC061233.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 AK143937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 AK172067.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 AK008096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 AK083630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 BC061233.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 AK143937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 AK172067.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214319 AK008096.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114163 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111253 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151319 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09601194 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09601215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09601190 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09601207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28013001 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28013004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28013005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09700140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09601190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09601204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09326243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013043 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10380299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380302 - -
          Affymetrix HT MG-430 PM Array Plate 1457846_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429188_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10380297 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389816 - -
          Mouse Genome 430 2.0 Array 1429188_at - -
          Mouse Genome 430 2.0 Array 1457846_at - -
          Mouse Genome 430 2.0 Array 1429188_PM_at - -
          Mouse Genome 430 2.0 Array 1457846_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69802_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69802_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100013088.mm.1 NM_199008.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013088.mm.1 NM_199008.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013088.mm.1 TC1100001400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013093.mm.1 NM_199008.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013093.mm.1 NM_199008.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013093.mm.1 TC1100001400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013095.mm.1 NM_199008.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013095.mm.1 NM_199008.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013095.mm.1 TC1100001400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013090.mm.1 NM_199008.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013090.mm.1 NM_199008.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013090.mm.1 TC1100001400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013092.mm.1 NM_199008.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013092.mm.1 NM_199008.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013092.mm.1 TC1100001400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013091.mm.1 NM_199008.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013091.mm.1 NM_199008.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013091.mm.1 TC1100001400.mm.1 MTA TC
          Mouse Expression Set 430 1457846_PM_at - -
          Mouse Expression Set 430 1429188_PM_at - -
          Mouse Expression Set 430 1457846_at - -
          Mouse Expression Set 430 1429188_at - -
          Murine Genome U74v2 Set 170663_PM_at - -
          Murine Genome U74v2 Set 170663_at - -
          Affymetrix HT Mouse Genome 430B Array 1429188_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457846_at - -
          Affymetrix HT Mouse Genome 430B Array 1457846_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429188_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1580121 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580121 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247397 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247397 - -
          mouse-6 v1.1 (Illumina) scl41046.4.1_69-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580121 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41046.4.1_69-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247397 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247397 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41046.4.1_69-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline