Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01617542_g1
          See other RPS16 GE Assays ›
          Gene Symbol:
          Rps16
          Gene Name:
          ribosomal protein S16
          Gene Aliases:
          AA420385, AI317031
          Chromosome Location:
          Chr.7: 28350689 - 28352698 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01617542_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013647.2 NP_038675.2 4 - 5 412 91
          GenBank mRNA AK002904.1 - 1 350 91
          AK010613.1 - 1 353 91
          AK010641.1 - 1 353 91
          AK011060.1 - 1 364 91
          AK019400.2 - 1 351 91
          AK083946.1 - 1 351 91
          AK088413.1 - 1 352 91
          AK088716.1 - 1 352 91
          AK150808.1 - 1 354 91
          AK168207.1 - 1 351 91
          AK169328.1 - 1 352 91
          BC082286.1 - 1 331 91
          BC090618.1 - 1 379 5713263 91
          BC115673.1 - 1 319 40104131 91
          BC115674.1 - 1 319 40104132 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps16

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          translation
          ribosomal small subunit biogenesis
          RNA binding
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021322 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021323 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021326 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021325 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021324 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963061 BC090618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963061 NM_013647.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2132844 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963062 AK150615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963062 AK150808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963062 AK168207.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963062 AK169328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963062 BC082286.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14229 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145347 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963062 AK150615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963062 AK150808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963062 AK168207.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963062 AK169328.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963062 BC082286.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2132844 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963061 BC090618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963061 NM_013647.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416995 AK150615.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416995 AK150808.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416995 AK168207.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416995 AK169328.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416995 BC082286.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021327 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145347 - -
          Whole Mouse Genome G4122A A_51_P416995 AK150615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416995 AK150808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416995 AK168207.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416995 AK169328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416995 BC082286.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106561 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06105446 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06105445 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06105467 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06105448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06105473 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021324 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021323 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021326 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021322 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06404224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06100637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06105467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06105470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06618987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06342349 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551491 - -
          Affymetrix HT MG-430 PM Array Plate 1455835_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551489 - -
          Affymetrix Mouse Gene 1.1 ST Array 10534053 - -
          Affymetrix Mouse Gene 1.1 ST Array 10578613 - -
          Mouse Genome 430 2.0 Array 1437196_x_at - -
          Mouse Genome 430 2.0 Array 1416404_PM_s_at - -
          Mouse Genome 430 2.0 Array 1455835_at - -
          Mouse Genome 430 2.0 Array 1416404_s_at - -
          Mouse Genome 430 2.0 Array 1455835_PM_at - -
          Mouse Genome 430 2.0 Array 1437196_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR0700004005.mm.1 TC0700000495.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004005.mm.1 NM_013647.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004005.mm.1 NM_013647.2 Refseq
          Mouse Expression Set 430 1416404_s_at - -
          Mouse Expression Set 430 1416404_PM_s_at - -
          Mouse Expression Set 430 1437196_PM_x_at - -
          Mouse Expression Set 430 1437196_x_at - -
          Mouse Expression Set 430 1455835_PM_at - -
          Mouse Expression Set 430 1455835_at - -
          Murine Genome U74v2 Set 97647_PM_at - -
          Murine Genome U74v2 Set 97647_at - -
          Murine Genome U74v2 Set 169860_PM_r_at - -
          Murine Genome U74v2 Set 169860_r_at - -
          Mouse Genome 430A 2.0 Array 1416404_s_at - -
          Mouse Genome 430A 2.0 Array 1416404_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1455835_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455835_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 904208 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 510722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692247 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692247 - -
          mouse-6 v1.1 (Illumina) scl020055.2_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 510722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020055.2_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227972 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227972 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020055.2_9-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline