Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01617724_g1
          See other CRXOS GE Assays ›
          Gene Symbol:
          Crxos
          Gene Name:
          cone-rod homeobox, opposite strand
          Gene Aliases:
          AA606869, AY590891, Crxos1, Egam1
          Chromosome Location:
          Chr.7: 15882617 - 15904030 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01617724_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crxos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033638.2 NP_001028810.1 4 - 5 606 116
          NM_001145190.2 NP_001138662.1 2 - 3 243 116
          GenBank mRNA AB472692.1 - 4 - 5 559 116
          AB472694.1 - 2 - 3 227 116
          AK145701.1 - 4 - 5 606 116
          AY590890.1 - 2 - 3 300 116
          AY590891.1 - 3 - 4 474 116
          CJ040988.1 - 2 - 3 226 116

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          helix-turn-helix transcription factor homeodomain transcription factor DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123237 C80347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123237 AK149251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123237 AK149256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P117786 NM_001033638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P117786 AK145701.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256696 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37467 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P117786 NM_001033638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P117786 AK145701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123237 C80347.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123237 AK149251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123237 AK149256.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09518402 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03807688 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09518386 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09518403 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09518385 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09518409 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09518406 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03807689 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09518388 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09518391 - -
          Whole Mouse Genome G4122A A_52_P485275 AK089116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P485275 NR_002167.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09518385 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09518403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875828 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550220 - -
          Affymetrix HT MG-430 PM Array Plate 1439973_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460605_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550217 - -
          Mouse Genome 430 2.0 Array 1439973_PM_at - -
          Mouse Genome 430 2.0 Array 1460605_PM_at - -
          Mouse Genome 430 2.0 Array 1460605_at - -
          Mouse Genome 430 2.0 Array 1439973_at - -
          Affymetrix Mouse Clariom S Array PSR0700001509.mm.1 TC0700000214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001509.mm.1 NM_001033638.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001509.mm.1 NM_001033638.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001509.mm.1 NM_001145190.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001509.mm.1 NM_001145190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001511.mm.1 TC0700000214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001511.mm.1 NM_001033638.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001511.mm.1 NM_001033638.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001511.mm.1 NM_001145190.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001511.mm.1 NM_001145190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001507.mm.1 TC0700000214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001507.mm.1 NM_001145190.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001507.mm.1 NM_001145190.2 Refseq
          Mouse Expression Set 430 1460605_at - -
          Mouse Expression Set 430 1439973_at - -
          Mouse Expression Set 430 1460605_PM_at - -
          Mouse Expression Set 430 1439973_PM_at - -
          Murine Genome U74v2 Set 95353_PM_at - -
          Murine Genome U74v2 Set 95353_at - -
          Mouse Genome 430A 2.0 Array 1460605_at - -
          Mouse Genome 430A 2.0 Array 1439973_PM_at - -
          Mouse Genome 430A 2.0 Array 1439973_at - -
          Mouse Genome 430A 2.0 Array 1460605_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439973_at - -
          Affymetrix HT Mouse Genome 430A Array 1460605_at - -
          Affymetrix HT Mouse Genome 430A Array 1439973_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460605_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893149 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3061023 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3138419 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3138419 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3061023 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline