Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01618442_m1
          See other GM5592 GE Assays ›
          Gene Symbol:
          Gm5592
          Gene Name:
          predicted gene 5592
          Gene Aliases:
          EG434172
          Chromosome Location:
          Chr.7: 41284327 - 41290183 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01618442_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5592
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033782.3 NP_001028954.1 2 - 3 909 152
          GenBank mRNA AK133295.1 - 2 - 3 910 152
          BC147594.1 - 2 - 3 794 9056584 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000727.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978621 NM_001033782.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978621 AK133295.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978621 NM_001033782.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978621 AK133295.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01503823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25076906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25076913 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552305 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552303 - -
          Affymetrix Mouse Clariom S Array PSR0700005919.mm.1 TC0700000727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005919.mm.1 NM_001033782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005919.mm.1 NM_001033782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005920.mm.1 TC0700000727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005920.mm.1 NM_001033782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005920.mm.1 NM_001033782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005918.mm.1 TC0700000727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005918.mm.1 NM_001033782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005918.mm.1 NM_001033782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005922.mm.1 TC0700000727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005922.mm.1 NM_001033782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005922.mm.1 NM_001033782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005921.mm.1 TC0700000727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005921.mm.1 NM_001033782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005921.mm.1 NM_001033782.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894054 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3032304 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3105802 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3032304 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3105802 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline