Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01618810_s1
          See other OLFR373 GE Assays ›
          Gene Symbol:
          Olfr373
          Gene Name:
          olfactory receptor 373
          Gene Aliases:
          MOR282-2
          Chromosome Location:
          Chr.8: 72099762 - 72100706 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01618810_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr373
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146539.2 NP_666750.2 1 610 179
          GenBank mRNA BC106979.2 - 1 608 179

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr373

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087567 BC150671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087567 NM_207555.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087567 BC119445.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087567 NM_146539.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087567 BC106979.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993298 BC119445.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993298 NM_146539.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993298 BC106979.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993298 BC106980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993298 BC119444.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087567 BC150671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087567 NM_207555.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087567 BC119445.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087567 NM_146539.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087567 BC106979.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993298 BC119445.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993298 NM_146539.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993298 BC106979.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993298 BC106980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993298 BC119444.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P476635 BC119445.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P476635 NM_146539.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P476635 BC106979.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P476635 BC106980.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P476635 BC119444.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959657 - -
          Whole Mouse Genome G4122A A_52_P476635 BC119445.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476635 NM_146539.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476635 BC106979.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476635 BC106980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476635 BC119444.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136371 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127168 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07394221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572742 - -
          Affymetrix Mouse Gene 1.1 ST Array 10572743 - -
          Affymetrix Mouse Gene 1.1 ST Array 10572741 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1450750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2663596 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2663596 - -
          mouse-6 v1.1 (Illumina) scl00258532.1_180-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450750 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258532.1_180-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450750 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2663595 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2663595 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline