Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01619091_m1
          See other RTP1 GE Assays ›
          Gene Symbol:
          Rtp1
          Gene Name:
          receptor transporter protein 1
          Gene Aliases:
          Gm1766, Gm604
          Chromosome Location:
          Chr.16: 23428601 - 23433960 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01619091_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rtp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004151.2 NP_001004151.1 1 - 2 318 131
          GenBank mRNA AK047543.1 - 1 - 2 319 131
          AY562225.1 - 1 - 2 272 131
          BC138910.1 - 1 - 2 304 8861932 131
          BC138912.1 - 1 - 2 304 8861934 131
          EU070411.1 - 1 - 2 164 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000343.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rtp1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein insertion into membrane
          olfactory receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30602962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30602963 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P415958 NM_001004151.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P415958 AK047543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P415958 NM_001004151.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P415958 AK047543.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665564 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156127 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P415958 NM_001004151.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P415958 AK047543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P415958 NM_001004151.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P415958 AK047543.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30602932 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156127 - -
          Whole Mouse Genome G4122A A_51_P415958 NM_001004151.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415958 AK047543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415958 NM_001004151.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415958 AK047543.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138488 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12682497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12682517 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12682510 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12682521 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12682526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30602962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30602963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30602903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12860580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12699825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13052764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12470701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12699829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12682497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12944807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12769141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12809617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12620966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12921155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12682510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12921152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12707751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12699823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12856459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12685231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12791247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12682514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12650628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12470697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30602953 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10434777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434776 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434774 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434775 - -
          Affymetrix Mouse Gene 1.1 ST Array 10434773 - -
          Affymetrix Mouse Clariom S Array PSR1600003409.mm.1 NM_001004151.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003409.mm.1 NM_001004151.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003409.mm.1 TC1600000343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003408.mm.1 NM_001004151.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003408.mm.1 NM_001004151.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003408.mm.1 TC1600000343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003410.mm.1 NM_001004151.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003410.mm.1 NM_001004151.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003410.mm.1 TC1600000343.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 477155 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224640 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2530019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2855737 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2855737 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline