Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01620193_s1
          See other TCEAL6 GE Assays ›
          Gene Symbol:
          Tceal6
          Gene Name:
          transcription elongation factor A (SII)-like 6
          Gene Aliases:
          1500026B10Rik
          Chromosome Location:
          Chr.X: 135208686 - 135210687 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01620193_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tceal6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025355.4 NP_079631.2 3 1046 73
          GenBank mRNA AK005297.1 - 3 1048 73
          BC087890.1 - 3 1046 6533845 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003007.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tceal6

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121288 NM_025355.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121288 AK005297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121288 BC087890.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121288 NM_025355.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121288 AK005297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121288 BC087890.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32675849 - -
          Whole Mouse Genome G4122A A_51_P349772 AK003156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349772 AK017585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349772 AK158996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349772 BC027131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349772 NM_001029978.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120757 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15080479 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15080483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15080490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15080478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15080494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32675798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14839646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32675819 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10601876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606790 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606789 - -
          Affymetrix Mouse Gene 1.1 ST Array 10601874 - -
          Affymetrix Mouse Clariom S Array PSR0X00018828.mm.1 NM_025355.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018828.mm.1 NM_025355.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018828.mm.1 TC0X00003007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018829.mm.1 NM_025355.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018829.mm.1 NM_025355.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018829.mm.1 TC0X00003007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018827.mm.1 NM_025355.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018827.mm.1 NM_025355.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018827.mm.1 TC0X00003007.mm.1 MTA TC
          Mouse Expression Set 430 1450961_a_at - -
          Mouse Expression Set 430 1450961_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450961_a_at - -
          Mouse Genome 430A 2.0 Array 1450961_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 576937 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2120577 - -
          Illumina mouse-6 v1.0 expression beadchip 540064 - -
          Illumina mouse-6 v1.0 expression beadchip 2120577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775324 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236300 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775324 - -
          mouse-6 v1.1 (Illumina) scl0066104.1_1-S - -
          mouse-6 v1.1 (Illumina) scl53824.3_522-S - -
          Illumina Sentrix mouse-6.v1 bead array 2120577 - -
          Illumina Sentrix mouse-6.v1 bead array 540064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53824.3_522-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066104.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120577 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2839027 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236300 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2839027 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775324 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0066104.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline