Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01621094_s1
          See other VMN2R121 GE Assays ›
          Gene Symbol:
          Vmn2r121
          Gene Name:
          vomeronasal 2, receptor 121
          Gene Aliases:
          EG625699
          Chromosome Location:
          Chr.X: 124127339 - 124135910 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01621094_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r121
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100616.1 NP_001094086.1 3 1041 131

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090152 NM_001100616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031547 NM_001100616.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513513 AK035593.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130942 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090152 NM_001100616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031547 NM_001100616.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32637754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14993373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14593717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32637770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373688 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606559 - -
          Affymetrix Mouse Clariom S Array PSR0X00018430.mm.1 NM_001100616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018430.mm.1 NM_001100616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018430.mm.1 TC0X00002931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018427.mm.1 NM_001100616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018427.mm.1 NM_001100616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018427.mm.1 TC0X00002931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018426.mm.1 NM_001100616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018426.mm.1 NM_001100616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018426.mm.1 TC0X00002931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018428.mm.1 NM_001100616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018428.mm.1 NM_001100616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018428.mm.1 TC0X00002931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018431.mm.1 NM_001100616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018431.mm.1 NM_001100616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018431.mm.1 TC0X00002931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018429.mm.1 NM_001100616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018429.mm.1 NM_001100616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018429.mm.1 TC0X00002931.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 660293 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline