Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01621922_g1
          See other RNF7 GE Assays ›
          Gene Symbol:
          Rnf7
          Gene Name:
          ring finger protein 7
          Gene Aliases:
          Rbx2, SAG
          Chromosome Location:
          Chr.9: 96470957 - 96478701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01621922_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011279.2 NP_035409.1 3 940 148
          GenBank mRNA AK002628.1 - 3 939 148
          AK003248.1 - 3 941 148
          AK003963.1 - 3 940 148
          AK011328.1 - 3 939 148
          AK146030.1 - 3 941 148
          AK208401.1 - 1 181 148
          BC011127.1 - 3 938 148

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of cysteine-type endopeptidase activity involved in apoptotic process
          intrinsic apoptotic signaling pathway in response to oxidative stress
          apoptotic mitochondrial changes
          cerebral cortex radially oriented cell migration
          radial glia guided migration of Purkinje cell
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          protein neddylation
          protein binding
          zinc ion binding
          NEDD8 transferase activity
          metal ion binding
          ubiquitin protein ligase activity
          cullin family protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26747279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26747280 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26747275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26747276 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003115 AK146030.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003115 AK011328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003115 AK209943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003115 BC011127.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003115 AK002628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065501 AK146030.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065501 AK208401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065501 AK011328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065501 BC011127.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065501 AK002628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110837 AK146030.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110837 AK011328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110837 BC011127.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110837 AK002628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110837 AK003248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352971 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 158379 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103932 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973521 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065501 AK146030.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065501 AK208401.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065501 AK011328.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065501 BC011127.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065501 AK002628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110837 AK146030.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110837 AK011328.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110837 BC011127.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110837 AK002628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110837 AK003248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003115 AK146030.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003115 AK011328.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003115 AK209943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003115 BC011127.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003115 AK002628.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414166 AF092877.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103932 - -
          Whole Mouse Genome G4122A A_51_P414166 AF092877.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43111 AK146030.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43111 AK011328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43111 BC011127.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43111 AK002628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43111 AK003248.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103932 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08072938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08019769 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26747276 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26747280 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26747275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26747279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26747193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08027194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07897523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08128820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07840595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08154753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07623874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07651330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07453037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07593999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08027187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07827827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07453040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07473696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07657648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07651328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07473693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07724579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26747318 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10595804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353009 - -
          Affymetrix HT MG-430 PM Array Plate 1426414_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1456600_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10595803 - -
          Mouse Genome 430 2.0 Array 1456600_PM_a_at - -
          Mouse Genome 430 2.0 Array 1456600_a_at - -
          Mouse Genome 430 2.0 Array 1447203_at - -
          Mouse Genome 430 2.0 Array 1426414_PM_a_at - -
          Mouse Genome 430 2.0 Array 1426414_a_at - -
          Mouse Genome 430 2.0 Array 1447203_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19823_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19823_at - -
          Affymetrix Mouse Clariom S Array PSR0900023709.mm.1 NM_011279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900023709.mm.1 TC0900002879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900023708.mm.1 NM_011279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900023708.mm.1 TC0900002879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900023713.mm.1 NM_011279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900023713.mm.1 TC0900002879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900023711.mm.1 NM_011279.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900023711.mm.1 TC0900002879.mm.1 MTA TC
          Mouse Expression Set 430 1426414_PM_a_at - -
          Mouse Expression Set 430 1426414_a_at - -
          Mouse Expression Set 430 1456600_PM_a_at - -
          Mouse Expression Set 430 1456600_a_at - -
          Murine Genome U74v2 Set 97836_PM_at - -
          Murine Genome U74v2 Set 97836_at - -
          Mouse Genome 430A 2.0 Array 1456600_a_at - -
          Mouse Genome 430A 2.0 Array 1456600_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426414_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426414_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1456600_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1456600_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426414_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426414_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 904330 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2754636 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2754636 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline