Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01622471_s1
          See other ACOT1 GE Assays ›
          Gene Symbol:
          Acot1
          Gene Name:
          acyl-CoA thioesterase 1
          Gene Aliases:
          ACH2, CTE-1, CTE-I, Cte1, D12Ucla1
          Chromosome Location:
          Chr.12: 84009502 - 84017670 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01622471_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acot1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012006.2 NP_036136.1 3 1478 94
          GenBank mRNA AK053680.1 - 1492 94
          BC003911.1 - 3 1475 94
          BC022951.1 - 2 2528 94
          BC028261.1 - 3 1488 5100478 94
          BC040749.1 - 2 2528 94
          Y14004.1 - 3 1478 94

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acot1

          Panther Classification:

          Molecular Function -

          esterase hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          long-chain fatty acid metabolic process
          acyl-CoA metabolic process
          palmitoyl-CoA hydrolase activity
          hydrolase activity
          thiolester hydrolase activity
          acyl-CoA hydrolase activity
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28595873 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038358 BC040749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038358 Y14004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038358 BC003911.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038358 BC022951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038358 BC028261.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79941 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147975 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038358 BC040749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038358 Y14004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038358 BC003911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038358 BC022951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038358 BC028261.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451574 BC040749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451574 Y14004.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451574 BC003911.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451574 BC022951.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451574 BC028261.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28595880 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02565122 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07689434 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147975 - -
          Whole Mouse Genome G4122A A_51_P451574 BC040749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451574 Y14004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451574 BC003911.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451574 BC022951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451574 BC028261.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137436 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147975 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776281 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776286 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776295 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28595873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28595905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10242976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09984233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09888843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10014169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09776286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09984234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09776289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10239577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10242971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10192099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595914 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10397149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397150 - -
          Affymetrix Mouse Gene 1.1 ST Array 10397148 - -
          Mouse Genome 430 2.0 Array 1449065_PM_at - -
          Mouse Genome 430 2.0 Array 1422997_s_at - -
          Mouse Genome 430 2.0 Array 1422997_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449065_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26897_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26897_PM_at - -
          Mouse Expression Set 430 1422997_s_at - -
          Mouse Expression Set 430 1449065_at - -
          Mouse Expression Set 430 1449065_PM_at - -
          Mouse Expression Set 430 1422997_PM_s_at - -
          Murine Genome U74v2 Set 103581_at - -
          Murine Genome U74v2 Set 103581_PM_at - -
          Mouse Genome 430A 2.0 Array 1449065_PM_at - -
          Mouse Genome 430A 2.0 Array 1449065_at - -
          Mouse Genome 430A 2.0 Array 1422997_s_at - -
          Mouse Genome 430A 2.0 Array 1422997_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4780600 - -
          Illumina mouse-6 v1.0 expression beadchip 4780600 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780600 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2768647 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2768647 - -
          mouse-6 v1.1 (Illumina) scl026897.4_185-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780600 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl026897.4_185-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780600 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3139875 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3062267 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2768647 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3139875 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3062267 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0