Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01622915_s1
          See other VMN2R104 GE Assays ›
          Gene Symbol:
          Vmn2r104
          Gene Name:
          vomeronasal 2, receptor 104
          Gene Aliases:
          V2r7
          Chromosome Location:
          Chr.17: 20029425 - 20048205 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01622915_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r104
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104566.1 NP_001098036.1 3 1195 137

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122614 NM_001104566.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113602 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122614 NM_001104566.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31081273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13579638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13250582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13579641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31081275 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448144 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442134 - -
          Affymetrix Mouse Gene 1.1 ST Array 10448141 - -
          Affymetrix Mouse Clariom S Array PSR1700013594.mm.1 NM_001104566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013594.mm.1 NM_001104566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013594.mm.1 TC1700001531.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013596.mm.1 NM_001104566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013596.mm.1 NM_001104566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013596.mm.1 TC1700001531.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013592.mm.1 NM_001104566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013592.mm.1 NM_001104566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013592.mm.1 TC1700001531.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013593.mm.1 NM_001104566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013593.mm.1 NM_001104566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013593.mm.1 TC1700001531.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013591.mm.1 NM_001104566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013591.mm.1 NM_001104566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013591.mm.1 TC1700001531.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013595.mm.1 NM_001104566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013595.mm.1 NM_001104566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013595.mm.1 TC1700001531.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 892162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline