| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700013685.mm.1 |
NM_001256493.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013685.mm.1 |
NM_001256493.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013685.mm.1 |
TC1700001561.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013682.mm.1 |
NM_001256493.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013682.mm.1 |
NM_001256493.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013682.mm.1 |
TC1700001561.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013684.mm.1 |
NM_001256493.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013684.mm.1 |
NM_001256493.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013684.mm.1 |
TC1700001561.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013679.mm.1 |
NM_001256493.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013679.mm.1 |
NM_001256493.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013679.mm.1 |
TC1700001561.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013677.mm.1 |
NM_001256493.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013677.mm.1 |
NM_001256493.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013677.mm.1 |
TC1700001561.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013678.mm.1 |
NM_001256493.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013678.mm.1 |
NM_001256493.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013678.mm.1 |
TC1700001561.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013683.mm.1 |
NM_001256493.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013683.mm.1 |
NM_001256493.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013683.mm.1 |
TC1700001561.mm.1 |
MTA TC |
| Murine Genome U74v2 Set |
171137_i_at |
- |
- |
| Murine Genome U74v2 Set |
171137_PM_i_at |
- |
- |