| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10442119 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442111 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442116 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442117 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442118 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10442115 |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
433070_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
433070_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1700001769.mm.1 |
NM_001104547.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001769.mm.1 |
NM_001104547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001769.mm.1 |
TC1700000223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001768.mm.1 |
NM_001104547.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001768.mm.1 |
NM_001104547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001768.mm.1 |
TC1700000223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001772.mm.1 |
NM_001104547.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001772.mm.1 |
NM_001104547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001772.mm.1 |
TC1700000223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001767.mm.1 |
NM_001104547.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001767.mm.1 |
NM_001104547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001767.mm.1 |
TC1700000223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001770.mm.1 |
NM_001104547.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001770.mm.1 |
NM_001104547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001770.mm.1 |
TC1700000223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001771.mm.1 |
NM_001104547.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001771.mm.1 |
NM_001104547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001771.mm.1 |
TC1700000223.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1420079_at |
- |
- |
| Mouse Expression Set 430 |
1420079_PM_at |
- |
- |
| Mouse Expression Set 430 |
1438124_at |
- |
- |
| Mouse Expression Set 430 |
1438124_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420079_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420079_PM_at |
- |
- |