Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01700131_s1
          See other TAS2R138 GE Assays ›
          Gene Symbol:
          Tas2r138
          Gene Name:
          taste receptor, type 2, member 138
          Gene Aliases:
          T2R138, Tas2r38, mt2r31
          Chromosome Location:
          Chr.6: 40612315 - 40613310 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01700131_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r138
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001451.1 NP_001001451.1 1 856 96
          GenBank mRNA AY916509.1 - 1 856 96
          BC128016.1 - 1 860 40136088 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r138

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity
          taste receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002908 BC128016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002908 NM_001001451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002908 AY916509.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100903 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002908 BC128016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002908 NM_001001451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002908 AY916509.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24389675 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100903 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05553564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05553568 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05553586 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05553581 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05553575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24389705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05060440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05605820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05646166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05605825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05251758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05553567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05837057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05553586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05605818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24389671 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10544285 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544284 - -
          Affymetrix Mouse Clariom S Array PSR0600016919.mm.1 TC0600002143.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016919.mm.1 NM_001001451.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016919.mm.1 NM_001001451.1 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162056 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162056 - -
          mouse-6 v1.1 (Illumina) GI_47824875-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline