Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01700139_s1
          See other TAS2R143 GE Assays ›
          Gene Symbol:
          Tas2r143
          Gene Name:
          taste receptor, type 2, member 143
          Gene Aliases:
          Gm1404, Tas2r43, mt2r36
          Chromosome Location:
          Chr.6: 42400238 - 42401119 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01700139_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r143
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001452.1 NP_001001452.1 1 766 114
          GenBank mRNA BC111103.1 - 1 765 114
          BC148239.1 - 1 766 40050382 114

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r143

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity
          taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P104119 NM_001001452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P104119 BC111103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P104119 BC148239.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P104119 NM_001001452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P104119 BC111103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P104119 BC148239.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398158 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104119 NM_001001452.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104119 BC111103.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104119 BC148239.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05158544 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05158535 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05280216 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05158543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05322838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05158542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05693823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05393850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05393851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05393853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05693827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05393848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05393862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05280215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05158529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537786 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537785 - -
          Affymetrix Mouse Clariom S Array PSR0600003721.mm.1 TC0600000461.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003721.mm.1 NM_001001452.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003721.mm.1 NM_001001452.1 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245879 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2897041 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2897041 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0