Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01700416_m1
          See other AVPR1B GE Assays ›
          Gene Symbol:
          Avpr1b
          Gene Name:
          arginine vasopressin receptor 1B
          Gene Aliases:
          AVPR3, V3/V1b, VIBR, VPR3
          Chromosome Location:
          Chr.1: 131599114 - 131612000 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01700416_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Avpr1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011924.2 NP_036054.1 1 - 2 1564 61
          GenBank mRNA AK033416.1 - 1 - 2 598 61
          AK050523.1 - 1 - 2 646 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001171.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Avpr1b

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure by vasopressin
          signal transduction
          G-protein coupled receptor signaling pathway
          cellular response to hormone stimulus
          positive regulation of vasoconstriction
          response to peptide
          signal transducer activity
          G-protein coupled receptor activity
          vasopressin receptor activity
          peptide hormone binding
          peptide binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121039 NM_011924.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121039 AK033416.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116610 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159401 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100691 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121039 NM_011924.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121039 AK033416.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P220454 AK050523.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P220454 NM_011924.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P220454 AK033416.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100691 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04264710 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159401 - -
          Whole Mouse Genome G4122A A_51_P220454 AK050523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220454 NM_011924.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220454 AK033416.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117732 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121039 NM_011924.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121039 AK033416.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100691 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159401 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00360279 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00916024 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00999801 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00916021 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00999795 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00916029 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00916023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00360280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00360277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20616694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00766265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01005314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00766262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00631659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00676342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00631654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00736162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00720950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00736161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00850538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00720955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20616752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00766260 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10349647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349645 - -
          Affymetrix HT MG-430 PM Array Plate 1422204_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10349644 - -
          Mouse Genome 430 2.0 Array 1422204_at - -
          Mouse Genome 430 2.0 Array 1422204_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26361_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26361_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100009438.mm.1 NM_011924.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009438.mm.1 NM_011924.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009438.mm.1 TC0100001171.mm.1 MTA TC
          Mouse Expression Set 430 1422204_PM_at - -
          Mouse Expression Set 430 1422204_at - -
          Mouse Genome 430A 2.0 Array 1422204_PM_at - -
          Mouse Genome 430A 2.0 Array 1422204_at - -
          Affymetrix HT Mouse Genome 430A Array 1422204_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422204_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 483492 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1190707 - -
          Illumina mouse-6 v1.0 expression beadchip 1190707 - -
          Illumina mouse-6 v1.0 expression beadchip 100510563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190707 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2667432 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2467247 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2667432 - -
          mouse-6 v1.1 (Illumina) scl0026361.2_34-S - -
          mouse-6 v1.1 (Illumina) scl17497.3_83-S - -
          Illumina Sentrix mouse-6.v1 bead array 100510563 - -
          Illumina Sentrix mouse-6.v1 bead array 1190707 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0026361.2_34-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100510563 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190707 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2667432 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2667432 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline