Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701269_s1
          See other 5930422O12RIK GE Assays ›
          Gene Symbol:
          5930422O12Rik
          Gene Name:
          RIKEN cDNA 5930422O12 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701269_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5930422O12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK031179.1 - 1 3146 121
          BB037066.2 - 1 579 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115696 BB037066.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115696 AK031179.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115696 BB037066.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115696 AK031179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P564286 AK031179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P564286 AK080418.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07199837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06981127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06981128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07199839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06683814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06878211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25768978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769012 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1457538_PM_at - -
          Mouse Genome 430 2.0 Array 1457538_at - -
          Mouse Genome 430 2.0 Array 1457538_PM_at - -
          Mouse Expression Set 430 1457538_PM_at - -
          Mouse Expression Set 430 1457538_at - -
          Affymetrix HT Mouse Genome 430B Array 1457538_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457538_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 633658 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline