Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701527_m1
          See other 4930583K01RIK GE Assays ›
          Gene Symbol:
          4930583K01Rik
          Gene Name:
          RIKEN cDNA 4930583K01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 118243907 - 118245551 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701527_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930583K01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027879.1 1 - 2 58 94
          GenBank mRNA AK015785.1 - 1 - 2 59 94
          AK016343.1 - 1 - 2 59 94

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK016343.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365044 NR_027879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365044 AK015785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365044 AK016343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365044 NR_027879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365044 AK015785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365044 AK016343.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126861 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365044 NR_027879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365044 AK015785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365044 AK016343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365044 NR_027879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365044 AK015785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365044 AK016343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365044 NR_027879.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365044 AK015785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365044 AK016343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365044 NR_027879.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365044 AK015785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365044 AK016343.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P06467483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25457448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25457443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05938920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25457446 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10556615 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556613 - -
          Affymetrix Mouse Clariom S Array PSR0700014079.mm.1 TC0700001693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700014079.mm.1 NR_027879.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700014079.mm.1 NR_027879.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700014082.mm.1 TC0700001693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700014082.mm.1 NR_027879.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700014082.mm.1 NR_027879.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 652596 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline