Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701690_s1
          See other DBPHT2 GE Assays ›
          Gene Symbol:
          Dbpht2
          Gene Name:
          DNA binding protein with his-thr domain
          Gene Aliases:
          Spot2
          Chromosome Location:
          Chr.12: 74297474 - 74300468 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701690_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dbpht2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198866.2 NP_942566.1 1 1827 64
          GenBank mRNA AF547221.1 - 1 842 64
          AK158661.1 - 1 236 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dbpht2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113946 AK139464.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113946 NM_198866.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726304 AK017171.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726304 AK168454.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39111 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156289 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113946 AK139464.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113946 NM_198866.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28542607 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156289 - -
          Whole Mouse Genome G4122A A_52_P656667 AF547221.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P656667 AK158661.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P656667 BC141071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P656667 NM_198866.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10300726 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10300745 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10300728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10300730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10300740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28542623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09820720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10221506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10187395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10362444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09879859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09820744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10362445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09820711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09831659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09820700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10221485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10221505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10300739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10221501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10221504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09879854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10078068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09879850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10187402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09964650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09820749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28542540 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10396473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396475 - -
          Affymetrix HT MG-430 PM Array Plate 1456998_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10396472 - -
          Mouse Genome 430 2.0 Array 1456998_at - -
          Mouse Genome 430 2.0 Array 1456998_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 386753_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 386753_at - -
          Affymetrix Mouse Clariom S Array PSR1200004556.mm.1 NM_198866.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004556.mm.1 NM_198866.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004556.mm.1 TC1200000661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200004557.mm.1 NM_198866.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004557.mm.1 NM_198866.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004557.mm.1 TC1200000661.mm.1 MTA TC
          Mouse Expression Set 430 1456998_PM_at - -
          Mouse Expression Set 430 1456998_at - -
          Murine Genome U74v2 Set 138207_f_at - -
          Murine Genome U74v2 Set 138207_PM_f_at - -
          Murine Genome U74v2 Set 131117_at - -
          Murine Genome U74v2 Set 131117_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456998_at - -
          Affymetrix HT Mouse Genome 430B Array 1456998_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2680114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2637413 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2637413 - -
          mouse-6 v1.1 (Illumina) scl0386753.1_250-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0386753.1_250-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2637413 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2637413 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0386753.1_250-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0