Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701728_s1
          See other TAS2R134 GE Assays ›
          Gene Symbol:
          Tas2r134
          Gene Name:
          taste receptor, type 2, member 134
          Gene Aliases:
          T2R134, Tas2r34
          Chromosome Location:
          Chr.2: 51627511 - 51628407 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701728_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r134
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199158.1 NP_954609.1 1 854 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r134

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity
          taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185875 NM_199158.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117158 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185875 NM_199158.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21214775 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117158 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112378 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117158 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01704181 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01704180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01704166 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01704170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21214801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01704176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01833600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02226202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01817234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02138525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02006755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01704166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01817238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01704181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01704172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02006752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01303392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21214818 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10472049 - -
          Affymetrix Mouse Gene 1.1 ST Array 10472047 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 70092 - -
          Illumina mouse-6 v1.0 expression beadchip 70092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247403 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247403 - -
          mouse-6 v1.1 (Illumina) scl0387511.1_285-S - -
          Illumina Sentrix mouse-6.v1 bead array 70092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0387511.1_285-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70092 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247403 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247403 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0