Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701895_s1
          See other MRGPRB1 GE Assays ›
          Gene Symbol:
          Mrgprb1
          Gene Name:
          MAS-related GPR, member B1
          Gene Aliases:
          MrgB1
          Chromosome Location:
          Chr.7: 48444113 - 48456342 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701895_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrgprb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_205810.4 NP_991379.3 2 214 93
          GenBank mRNA AK148044.1 - 2 210 93
          AK148046.1 - 2 119 93
          AK148052.1 - 2 118 93
          AK148074.1 - 2 215 93
          AK148108.1 - 2 119 93
          AK148114.1 - 2 215 93
          AK148166.1 - 2 119 93
          AK148241.1 - 2 119 93
          AK148253.1 - 2 210 93
          AK148256.1 - 2 210 93
          AK148273.1 - 2 187 93
          AK148279.1 - 2 168 93
          AK148322.1 - 2 152 93
          AK148388.1 - 2 119 93
          BC116395.1 - 1 40 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrgprb1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          signal transducer activity
          G-protein coupled receptor activity
          neuropeptide binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 AK148388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 BC116395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 BC116396.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 AK148273.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 AK148279.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 AK148388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 BC116395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 BC116396.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 AK148273.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P288260 AK148279.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071940 AK148279.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071940 NM_205810.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645263 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81881 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141701 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103328 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071940 AK148279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071940 NM_205810.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 AK148388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 BC116395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 BC116396.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 AK148273.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 AK148279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 AK148388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 BC116395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 BC116396.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 AK148273.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P288260 AK148279.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 AK148388.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 BC116395.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 BC116396.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 AK148273.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 AK148279.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 AK148388.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 BC116395.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 BC116396.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 AK148273.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288260 AK148279.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25107281 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141701 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06189624 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103328 - -
          Whole Mouse Genome G4122A A_52_P288260 AK148388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 BC116395.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 BC116396.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 AK148273.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 AK148279.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 AK148388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 BC116395.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 BC116396.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 AK148273.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288260 AK148279.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128106 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103328 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25107308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06648416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06648409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06086757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05917309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06056068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06546661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06546663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25107268 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563751 - -
          Affymetrix HT MG-430 PM Array Plate 1437529_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563749 - -
          Mouse Genome 430 2.0 Array 1437529_PM_at - -
          Mouse Genome 430 2.0 Array 1437529_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233231_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233231_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700028000.mm.1 NM_205810.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028000.mm.1 NM_205810.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028000.mm.1 TC0700003068.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027999.mm.1 NM_205810.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027999.mm.1 NM_205810.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027999.mm.1 TC0700003068.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700028002.mm.1 NM_205810.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028002.mm.1 NM_205810.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028002.mm.1 TC0700003068.mm.1 MTA TC
          Mouse Expression Set 430 1437529_PM_at - -
          Mouse Expression Set 430 1437529_at - -
          Affymetrix HT Mouse Genome 430B Array 1437529_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437529_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4610154 - -
          Illumina mouse-6 v1.0 expression beadchip 4610154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004610154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245063 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245063 - -
          mouse-6 v1.1 (Illumina) scl0233231.1_107-S - -
          Illumina Sentrix mouse-6.v1 bead array 4610154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0233231.1_107-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4610154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245063 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245063 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0