Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701924_s1
          See other TLR11 GE Assays ›
          Gene Symbol:
          Tlr11
          Gene Name:
          toll-like receptor 11
          Gene Aliases:
          Gm287
          Chromosome Location:
          Chr.14: 50357914 - 50363663 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701924_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tlr11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_205819.3 NP_991388.2 2 2913 74
          GenBank mRNA AY510704.1 - 1 2868 74
          BC109164.2 - 1 2855 40048069 74
          BC109165.1 - 1 2853 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tlr11

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          signal transduction
          response to bacterium
          toll-like receptor 11 signaling pathway
          innate immune response
          regulation of cytokine secretion
          positive regulation of inflammatory response
          transmembrane signaling receptor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048289 NM_205819.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048289 AI646605.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243092 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171146 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048289 NM_205819.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048289 AI646605.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597150 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171146 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105552 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171146 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11427007 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11426969 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11426989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11217608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11396070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11426998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11180402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11180406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11426978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11178848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11759198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11427007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11426964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11759199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11426969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11426997 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414472 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414470 - -
          Affymetrix Mouse Clariom S Array PSR1400004416.mm.1 NM_205819.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004416.mm.1 TC1400000552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004411.mm.1 NM_205819.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004411.mm.1 TC1400000552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004414.mm.1 NM_205819.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004414.mm.1 TC1400000552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004413.mm.1 TC1400000552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004415.mm.1 NM_205819.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004415.mm.1 TC1400000552.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550673 - -
          Illumina mouse-6 v1.0 expression beadchip 5550673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605068 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605068 - -
          mouse-6 v1.1 (Illumina) scl0239081.1_288-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0239081.1_288-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2795644 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2795644 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline