Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01702116_s1
          See other OLFR1308 GE Assays ›
          Gene Symbol:
          Olfr1308
          Gene Name:
          olfactory receptor 1308
          Gene Aliases:
          MOR245-22
          Chromosome Location:
          Chr.2: 111960109 - 111961071 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01702116_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1308
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207151.1 NP_997034.1 1 889 93
          GenBank mRNA BC108948.2 - 1 887 93
          BC108949.2 - 1 888 93
          BC115847.1 - 1 888 93
          BC115949.1 - 1 888 40043972 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1308

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 NM_207151.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 BC108948.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 BC108949.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 BC115847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 BC115949.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 NM_207151.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 BC108948.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 BC108949.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 BC115847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P468150 BC115949.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179662 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 NM_207151.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 BC108948.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 BC108949.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 BC115847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 BC115949.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 NM_207151.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 BC108948.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 BC108949.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 BC115847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P468150 BC115949.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 NM_207151.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 BC108948.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 BC108949.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 BC115847.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 BC115949.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 NM_207151.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 BC108948.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 BC108949.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 BC115847.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468150 BC115949.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526870 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179662 - -
          Whole Mouse Genome G4122A A_51_P468150 NM_207151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 BC108948.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 BC108949.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 BC115847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 BC115949.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 NM_207151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 BC108948.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 BC108949.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 BC115847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468150 BC115949.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02122286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01905008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02122275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485801 - -
          Affymetrix Mouse Gene 1.1 ST Array 10485800 - -
          Affymetrix Mouse Clariom S Array PSR0200036238.mm.1 NM_207151.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036238.mm.1 NM_207151.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036238.mm.1 TC0200004336.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1500551 - -
          Illumina mouse-6 v1.0 expression beadchip 1500551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640178 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2640178 - -
          mouse-6 v1.1 (Illumina) scl0258258.1_83-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258258.1_83-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2640178 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2640178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0