Search
Search

Haglr| Gene Symbol: Haglr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110445.1 | 3 | 2403 | 97 | ||
| GenBank mRNA | AK032721.1 | - | 1 | 2038 | 97 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | NM_207266.1 | - | 2038 | Chr. - - [-] on Build | 97 | ||
Set Membership: |
|