Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01702510_sH
          See other OLFR1198 GE Assays ›
          Gene Symbol:
          Olfr1198
          Gene Name:
          olfactory receptor 1198
          Gene Aliases:
          GA_x5J8B7W62NC-67655-66851, GA_x6K02T2Q125-50221692-50220766, MOR225-13
          Chromosome Location:
          Chr.2: 88745960 - 88746886 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01702510_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1198
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207567.1 NP_997450.1 1 228 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004026.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1198

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237055 NM_207567.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237055 NM_207567.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237055 NM_207567.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237055 NM_207567.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P237055 NM_207567.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P237055 NM_207567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237055 NM_207567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237055 NM_207567.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115581 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01868148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01589980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02069659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01589978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01252659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01814854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01868146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01995129 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484759 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484758 - -
          Affymetrix Mouse Clariom S Array PSR0200033967.mm.1 NM_207567.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033967.mm.1 NM_207567.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033967.mm.1 TC0200004026.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 517162 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6290176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745761 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745761 - -
          mouse-6 v1.1 (Illumina) scl0404330.1_63-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0404330.1_63-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2745761 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2745761 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline