Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01702733_g1
          See other MUP5 GE Assays ›
          Gene Symbol:
          Mup5
          Gene Name:
          major urinary protein 5
          Gene Aliases:
          Mup18
          Chromosome Location:
          Chr.4: 61831319 - 61835180 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01702733_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mup5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008649.2 NP_032675.2 5 - 6 575 170
          GenBank mRNA M16360.1 - 5 - 6 575 170

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mup5

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          molecular_function
          transporter activity
          pheromone binding
          small molecule binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P412506 M16360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P412506 BC132310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P412506 BC132312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P412506 M16360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P412506 BC132310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P412506 BC132312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P412506 M16360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P412506 BC132310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P412506 BC132312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P412506 M16360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P412506 BC132310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P412506 BC132312.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22962245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22962256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22962265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22962262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22962272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22962243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22962244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22962261 - -
          Whole Mouse Genome G4122A A_52_P412506 M16360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P412506 BC132310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P412506 BC132312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P412506 M16360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P412506 BC132310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P412506 BC132312.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106531 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05256336 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03310335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03590334 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03904502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22962245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22962272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22962287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22962244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22962285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22962280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22962265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03526333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22962252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22962219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22962245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22962272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22962244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03441647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22962243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22962256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03727405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22962220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03310335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03441623 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10513469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513518 - -
          Affymetrix HT MG-430 PM Array Plate 1426166_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10513514 - -
          Mouse Genome 430 2.0 Array 1426166_PM_at - -
          Mouse Genome 430 2.0 Array 1426166_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17844_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17844_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400022699.mm.1 TC0400002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022699.mm.1 NM_008649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022699.mm.1 NM_008649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022701.mm.1 TC0400002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022701.mm.1 NM_008649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022701.mm.1 NM_008649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022696.mm.1 TC0400002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022696.mm.1 NM_008649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022696.mm.1 NM_008649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022702.mm.1 TC0400002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022702.mm.1 NM_008649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022702.mm.1 NM_008649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022703.mm.1 TC0400002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022703.mm.1 NM_008649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022703.mm.1 NM_008649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022697.mm.1 TC0400002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022697.mm.1 NM_008649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022697.mm.1 NM_008649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022698.mm.1 TC0400002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022698.mm.1 NM_008649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022698.mm.1 NM_008649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022700.mm.1 TC0400002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022700.mm.1 NM_008649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022700.mm.1 NM_008649.2 Refseq
          Mouse Expression Set 430 1426166_at - -
          Mouse Expression Set 430 1426166_PM_at - -
          Murine Genome U74v2 Set 101635_PM_f_at - -
          Murine Genome U74v2 Set 101635_f_at - -
          Murine Genome U74v2 Set 137167_PM_at - -
          Murine Genome U74v2 Set 137167_at - -
          Mouse Genome 430A 2.0 Array 1426166_at - -
          Mouse Genome 430A 2.0 Array 1426166_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426166_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426166_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 389200 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6770279 - -
          Illumina mouse-6 v1.0 expression beadchip 6770279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226884 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226884 - -
          mouse-6 v1.1 (Illumina) scl0017844.1_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017844.1_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3158509 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3079257 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226884 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226884 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3079257 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3158509 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline