Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01702786_gH
          See other VCP GE Assays ›
          Gene Symbol:
          Vcp
          Gene Name:
          valosin containing protein
          Gene Aliases:
          3110001E05, CDC48, p97, p97/VCP
          Chromosome Location:
          Chr.4: 42979964 - 43000507 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01702786_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vcp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009503.4 NP_033529.3 13 - 14 2024 115
          GenBank mRNA AK010612.1 - 3 - 4 374 115
          AK028264.1 - 13 - 14 1953 115
          AK030751.1 - 13 - 14 1855 115
          AK083821.1 - 11 - 12 1530 115
          AK146109.1 - 13 - 14 1919 115
          AK149931.1 - 13 - 14 1917 115
          AK151109.1 - 13 - 14 1918 115
          AK151418.1 - 13 - 14 1918 115
          AK153012.1 - 13 - 14 1955 115
          AK153249.1 - 13 - 14 1995 115
          AK159177.1 - 13 - 14 1953 115
          AK159475.1 - 13 - 14 1951 115
          AK159509.1 - 13 - 14 1996 115
          AK160889.1 - 13 - 14 1839 115
          AK167794.1 - 13 - 14 1972 115
          AK169140.1 - 13 - 14 1926 115
          BC043053.1 - 13 - 14 1928 6418662 115
          BC049114.1 - 13 - 14 1972 6810751 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vcp

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          double-strand break repair
          ubiquitin-dependent protein catabolic process
          transport
          ER to Golgi vesicle-mediated transport
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to DNA damage stimulus
          protein ubiquitination
          protein N-linked glycosylation via asparagine
          viral genome replication
          translesion synthesis
          ER-associated ubiquitin-dependent protein catabolic process
          retrograde protein transport, ER to cytosol
          positive regulation of protein complex assembly
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein hexamerization
          positive regulation of protein catabolic process
          protein homooligomerization
          aggresome assembly
          ER-associated misfolded protein catabolic process
          positive regulation of protein K63-linked deubiquitination
          positive regulation of Lys63-specific deubiquitinase activity
          nucleotide binding
          receptor binding
          protein binding
          ATP binding
          lipid binding
          hydrolase activity
          ATPase activity
          protein phosphatase binding
          protein domain specific binding
          polyubiquitin binding
          protein complex binding
          deubiquitinase activator activity
          identical protein binding
          ADP binding
          ubiquitin-like protein ligase binding
          poly(A) RNA binding
          ubiquitin-specific protease binding

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 901458 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline