Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01702813_m1
          See other KLRA3 GE Assays ›
          Gene Symbol:
          Klra3
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 3
          Gene Aliases:
          5E6, Ly49c, NK-2.1, Nk-2, Nk2, Nk2.1
          Chromosome Location:
          Chr.6: 130323289 - 130337626 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01702813_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289604.1 NP_001276533.1 2 - 3 287 106
          NM_001289605.1 NP_001276534.1 2 - 3 321 106
          NM_010648.3 NP_034778.2 2 - 3 287 106
          GenBank mRNA AK159595.1 - 2 - 3 236 106
          BC099867.1 - 119 40040425 106
          BC099878.1 - 119 40040424 106
          BC099879.1 - 119 40040423 106
          BC099880.1 - 119 40040422 106
          BC101952.1 - 2 - 3 163 40044699 106
          BC103543.1 - 2 - 3 163 40044703 106
          BC103544.1 - 2 - 3 197 40044701 106
          BC103545.1 - 2 - 3 163 40044700 106
          BC132524.1 - 2 - 3 168 40130801 106
          BC132526.1 - 2 - 3 168 40130803 106
          BC145192.1 - 2 - 3 168 9053743 106
          U09739.1 - 2 - 3 287 106
          U10305.1 - 2 - 3 203 106
          U49868.1 - 2 - 3 268 106
          U56404.1 - 2 - 3 162 106
          U56405.1 - 2 - 3 162 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003613.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra3

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468313 U09739.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468313 U10305.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468313 U49868.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468313 U56405.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468313 BC103544.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468308 U10305.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468308 AK159595.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468308 NM_010648.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P34058 U09739.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P34058 U10305.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P34058 U49868.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P34058 U56404.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P34058 U56405.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24847634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24847658 - -
          Whole Mouse Genome G4122A A_52_P34058 U09739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P34058 U10305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P34058 U49868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P34058 U56404.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P34058 U56405.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468308 U10305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468308 AK159595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468308 NM_010648.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468313 U09739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468313 U10305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468313 U49868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468313 U56405.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468313 BC103544.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05277331 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05862642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24847642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05862642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24847607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05376336 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548538 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548536 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548525 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548532 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548513 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548535 - -
          Mouse Genome 430 2.0 Array 1425436_PM_x_at - -
          Mouse Genome 430 2.0 Array 1425436_x_at - -
          Affymetrix Mouse Clariom S Array PSR0600025611.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025611.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025611.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025611.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025613.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025613.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025613.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025613.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025617.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025617.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025617.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025617.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025614.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025614.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025614.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025614.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025620.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025620.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025620.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025620.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025603.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025603.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025603.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025603.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025606.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025606.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025610.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025610.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025610.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025610.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025616.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025616.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025612.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025612.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025612.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025612.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025609.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025609.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025609.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025609.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025619.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025619.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025619.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025619.mm.1 NM_010648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025605.mm.1 TC0600003613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025605.mm.1 NM_001289604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025605.mm.1 NM_001289605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025605.mm.1 NM_010648.3 Refseq
          Mouse Expression Set 430 1425436_PM_x_at - -
          Mouse Expression Set 430 1425436_x_at - -
          Murine Genome U74v2 Set 93893_PM_f_at - -
          Murine Genome U74v2 Set 94779_f_at - -
          Murine Genome U74v2 Set 93894_PM_f_at - -
          Murine Genome U74v2 Set 93893_f_at - -
          Murine Genome U74v2 Set 94779_PM_f_at - -
          Murine Genome U74v2 Set 93894_f_at - -
          Mouse Genome 430A 2.0 Array 1425436_x_at - -
          Mouse Genome 430A 2.0 Array 1425436_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 857174 - -
          ABI Mouse Genome Survey Microarray 858328 - -
          ABI Mouse Genome Survey Microarray 915889 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1980097 - -
          MouseWG-6 v2 Expression BeadChip 101770048 - -
          Illumina mouse-6 v1.0 expression beadchip 101770048 - -
          Illumina mouse-6 v1.0 expression beadchip 1980097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2602871 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237580 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2602871 - -
          mouse-6 v1.1 (Illumina) scl0016634.1_61-S - -
          mouse-6 v1.1 (Illumina) GI_21426856-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980097 - -
          Illumina Sentrix mouse-6.v1 bead array 101770048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0016634.1_61-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101770048 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980097 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2716458 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237580 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0