Search
Search

Olfr638| Gene Symbol: Olfr638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147120.1 | NP_667331.1 | 1 | 44 | 163 | |
| GenBank mRNA | BC128011.1 | - | 1 | 43 | 40136021 | 163 |
| BC152352.1 | - | 1 | 48 | 40136020 | 163 | |
Set Membership: |
|