Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01703264_g1
          See other ELOF1 GE Assays ›
          Gene Symbol:
          Elof1
          Gene Name:
          elongation factor 1 homolog (ELF1, S. cerevisiae)
          Gene Aliases:
          1110011K10Rik
          Chromosome Location:
          Chr.9: 22112989 - 22114169 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01703264_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Elof1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_170777.3 NP_740747.1 1 - 2 363 179
          GenBank mRNA AK002505.1 - 197 179
          AK003614.1 - 201 179
          AK014116.1 - 177 179
          AK089052.1 - 187 179
          BC019870.1 - 170 5123727 179
          BC024488.1 - 182 4981209 179
          BC056225.1 - 1 - 2 363 5150539 179

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Elof1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          molecular_function
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350533 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350532 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 BC024488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 BC056225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 AK002505.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 AK003614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 NM_170777.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 BC024488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 BC056225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 AK002505.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 AK003614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158772 NM_170777.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566080 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137602 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 BC024488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 BC056225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 AK002505.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 AK003614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 NM_170777.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 BC024488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 BC056225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 AK002505.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 AK003614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158772 NM_170777.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 BC024488.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 BC056225.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 AK002505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 AK003614.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 NM_170777.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 BC024488.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 BC056225.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 AK002505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 AK003614.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158772 NM_170777.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350511 - -
          Whole Mouse Genome G4122A A_51_P158772 BC024488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 BC056225.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 AK002505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 AK003614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 NM_170777.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 BC024488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 BC056225.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 AK002505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 AK003614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158772 NM_170777.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108575 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07571626 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07705817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07686221 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07705818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07705806 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07571630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07571627 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350532 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07535470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08034919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07535469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07571627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07921983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07588321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08166577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07705804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07571630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07670020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07705818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07972550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07472655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350516 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591737 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591736 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591738 - -
          Affymetrix HT MG-430 PM Array Plate 1423820_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10591735 - -
          Mouse Genome 430 2.0 Array 1423820_at - -
          Mouse Genome 430 2.0 Array 1423820_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66126_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66126_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900016021.mm.1 NM_170777.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016021.mm.1 NM_170777.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016021.mm.1 NR_106052.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016021.mm.1 TC0900001899.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900016022.mm.1 NM_170777.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016022.mm.1 NM_170777.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016022.mm.1 TC0900001899.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900016019.mm.1 NM_170777.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016019.mm.1 NM_170777.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016019.mm.1 TC0900001899.mm.1 MTA TC
          Mouse Expression Set 430 1423820_PM_at - -
          Mouse Expression Set 430 1423820_at - -
          Murine Genome U74v2 Set 95760_PM_at - -
          Murine Genome U74v2 Set 95760_at - -
          Murine Genome U74v2 Set 170333_at - -
          Murine Genome U74v2 Set 170864_i_at - -
          Murine Genome U74v2 Set 170333_PM_at - -
          Murine Genome U74v2 Set 170864_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1423820_PM_at - -
          Mouse Genome 430A 2.0 Array 1423820_at - -
          Affymetrix HT Mouse Genome 430A Array 1423820_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423820_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 580019 - -
          Illumina mouse-6 v1.0 expression beadchip 580019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000580019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231371 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231371 - -
          mouse-6 v1.1 (Illumina) scl36121.3_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 580019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36121.3_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 580019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231371 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231371 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36121.3_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline