Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01703321_mH
          See other CYP3A44 GE Assays ›
          Gene Symbol:
          Cyp3a44
          Gene Name:
          cytochrome P450, family 3, subfamily a, polypeptide 44
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 145773983 - 145805891 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01703321_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp3a44
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177380.3 NP_796354.1 11 - 12 1344 102
          GenBank mRNA AB039380.1 - 11 - 12 1344 102
          BC125522.1 - 11 - 12 1306 40130037 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003599.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp3a44

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          demethylation
          oxidoreductase activity
          demethylase activity
          testosterone 6-beta-hydroxylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366803 BC125522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366803 NM_177380.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366803 AB039380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366803 BC125522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366803 NM_177380.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366803 AB039380.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122786 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366803 BC125522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366803 NM_177380.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366803 AB039380.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366803 BC125522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366803 NM_177380.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366803 AB039380.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366803 BC125522.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366803 NM_177380.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366803 AB039380.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366803 BC125522.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366803 NM_177380.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P366803 AB039380.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P571006 BC125522.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P571006 NM_177380.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P571006 AB039380.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166075 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05779961 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05779974 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05779931 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05779941 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05779927 - -
          Whole Mouse Genome G4122A A_52_P571006 BC125522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P571006 NM_177380.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P571006 AB039380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366803 BC125522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366803 NM_177380.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366803 AB039380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366803 BC125522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366803 NM_177380.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366803 AB039380.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05779931 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05779927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04892383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04398395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04442916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04296289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166054 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10535678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535699 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535680 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535702 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535701 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535681 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535696 - -
          Affymetrix HT MG-430 PM Array Plate 1426064_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10535698 - -
          Affymetrix Mouse Gene 1.1 ST Array 10535683 - -
          Affymetrix Mouse Gene 1.1 ST Array 10535669 - -
          Mouse Genome 430 2.0 Array 1426064_PM_at - -
          Mouse Genome 430 2.0 Array 1426064_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 337924_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 337924_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500033033.mm.1 TC0500003599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033033.mm.1 NM_177380.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033033.mm.1 NM_177380.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033032.mm.1 TC0500003599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033032.mm.1 NM_177380.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033032.mm.1 NM_177380.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033028.mm.1 TC0500003599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033028.mm.1 NM_177380.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033028.mm.1 NM_177380.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033031.mm.1 TC0500003599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033031.mm.1 NM_177380.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033031.mm.1 NM_177380.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033029.mm.1 TC0500003599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033029.mm.1 NM_177380.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033029.mm.1 NM_177380.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033030.mm.1 TC0500003599.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033030.mm.1 NM_177380.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033030.mm.1 NM_177380.3 Refseq
          Mouse Expression Set 430 1426064_at - -
          Mouse Expression Set 430 1426064_PM_at - -
          Mouse Genome 430A 2.0 Array 1426064_PM_at - -
          Mouse Genome 430A 2.0 Array 1426064_at - -
          Affymetrix HT Mouse Genome 430A Array 1426064_at - -
          Affymetrix HT Mouse Genome 430A Array 1426064_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 657690 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2810717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002810717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247769 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247769 - -
          mouse-6 v1.1 (Illumina) scl00337924.1_318-S - -
          Illumina Sentrix mouse-6.v1 bead array 2810717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00337924.1_318-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2810717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247769 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247769 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline