Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01703405_m1
          See other CD300C GE Assays ›
          Gene Symbol:
          Cd300c
          Gene Name:
          CD300C antigen
          Gene Aliases:
          Clm6
          Chromosome Location:
          Chr.11: 114956278 - 114960417 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01703405_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd300c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199225.1 NP_954695.1 3 - 5 543 153
          GenBank mRNA AY457052.1 - 3 - 5 543 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004052.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus Inventoried

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd300c

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 422495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178869 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176856 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023562 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P23550 NM_199225.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P23550 AY457052.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150646 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176856 - -
          Whole Mouse Genome G4122A A_52_P23550 NM_199225.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P23550 AY457052.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09258601 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09258607 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09258602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09260451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09117208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09619006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09230574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09117206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09616627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09258609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09286757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09260454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09258606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09117207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09230570 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10392806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392807 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392805 - -
          Affymetrix Mouse Gene 1.1 ST Array 10392803 - -
          Affymetrix Mouse Clariom S Array PSR1100038009.mm.1 NM_199225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038009.mm.1 NM_199225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038009.mm.1 TC1100004052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038008.mm.1 NM_199225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038008.mm.1 NM_199225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038008.mm.1 TC1100004052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038016.mm.1 NM_199225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038016.mm.1 NM_199225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038016.mm.1 TC1100004052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038021.mm.1 NM_199225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038021.mm.1 NM_199225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038021.mm.1 TC1100004052.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5360072 - -
          Illumina mouse-6 v1.0 expression beadchip 5360072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216280 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216280 - -
          mouse-6 v1.1 (Illumina) scl0387565.3_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0387565.3_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216280 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216280 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline